missing translation for 'onlineSavingsMsg'
Learn More

4-(Hydroxymethyl)phenylacetic acid, 97%

Catalog No. AAH2753103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2753103 1 g
AAH2753106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2753103 Supplier Thermo Scientific Chemicals Supplier No. H2753103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 73401-74-8 | C9H10O3 | 166.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 73401-74-8
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00065692
InChI Key FWZBPBKAANKOJQ-UHFFFAOYSA-N
Synonym 4-hydroxymethyl phenylacetic acid, 2-4-hydroxymethyl phenyl acetic acid, 4-hydroxymethyl phenyl acetic acid, 4-hydroxymethylphenylacetic acid, benzeneacetic acid,4-hydroxymethyl, benzeneacetic acid, 4-hydroxymethyl, ambotzrl-1037, pubchem19630, acmc-1bl5i
PubChem CID 3283498
IUPAC Name 2-[4-(hydroxymethyl)phenyl]acetic acid
SMILES C1=CC(=CC=C1CC(=O)O)CO

Specifications

Melting Point 131°C to 134°C
Quantity 1 g
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material 4-(Hydroxymethyl)phenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only