missing translation for 'onlineSavingsMsg'
Learn More

4-(Hydroxymethyl)benzeneboronic acid, 98%

CAS: 59016-93-2 | C7H9BO3 | 151.956 g/mol

Supplier:  Thermo Scientific Chemicals L1519406

 View more versions of this product

Catalog No. AAL1519406

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-(Hydroxymethyl)benzeneboronic acid
∼280°C
MFCD00792672
2831413
PZRPBPMLSSNFOM-UHFFFAOYSA-N
[4-(hydroxymethyl)phenyl]boronic acid
2734706
98%
59016-93-2
C7H9BO3
5 g
4-hydroxymethyl phenylboronic acid, 4-hydroxymethyl benzeneboronic acid, 4-hydroxymethyl phenyl boronic acid, 4-hydroxymethylphenyl boronic acid, 4-hydroxymethylphenylboronic acid, 4-boronobenzyl alcohol, p-hydroxymethyl phenylboronic acid, 4-hydroxymethyl phenyl boranediol
B(C1=CC=C(C=C1)CO)(O)O
151.956
151.96

Chemical Identifiers

59016-93-2
151.956
PZRPBPMLSSNFOM-UHFFFAOYSA-N
2734706
B(C1=CC=C(C=C1)CO)(O)O
C7H9BO3
MFCD00792672
4-hydroxymethyl phenylboronic acid, 4-hydroxymethyl benzeneboronic acid, 4-hydroxymethyl phenyl boronic acid, 4-hydroxymethylphenyl boronic acid, 4-hydroxymethylphenylboronic acid, 4-boronobenzyl alcohol, p-hydroxymethyl phenylboronic acid, 4-hydroxymethyl phenyl boranediol
[4-(hydroxymethyl)phenyl]boronic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only