missing translation for 'onlineSavingsMsg'
Learn More

4-(Hydroxymethyl)benzeneboronic acid, 98%

Catalog No. aal1519403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1519403 1 g
AAL1519406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1519403 Supplier Thermo Scientific Chemicals Supplier No. L1519403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 59016-93-2 | C7H9BO3 | 151.956 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 59016-93-2
Molecular Formula C7H9BO3
Molecular Weight (g/mol) 151.956
MDL Number MFCD00792672
InChI Key PZRPBPMLSSNFOM-UHFFFAOYSA-N
Synonym 4-hydroxymethyl phenylboronic acid, 4-hydroxymethyl benzeneboronic acid, 4-hydroxymethyl phenyl boronic acid, 4-hydroxymethylphenyl boronic acid, 4-hydroxymethylphenylboronic acid, 4-boronobenzyl alcohol, p-hydroxymethyl phenylboronic acid, 4-hydroxymethyl phenyl boranediol
PubChem CID 2734706
IUPAC Name [4-(hydroxymethyl)phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)CO)(O)O

Specifications

Melting Point ∼280°C
Quantity 1 g
Beilstein 2831413
Formula Weight 151.96
Percent Purity 98%
Chemical Name or Material 4-(Hydroxymethyl)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only