missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxymethyl-4'-methyl-2,2'-bipyridyl 97.0+%, TCI America™
Supplier: TCI America H13471G
Specifications
| 4-Hydroxymethyl-4′-methyl-2,2′-bipyridyl | |
| 121°C | |
| C12H12N2O | |
| 1 g | |
| GJCOKGHYIMLMPB-UHFFFAOYSA-N | |
| {4'-methyl-[2,2'-bipyridin]-4-yl}methanol | |
| 2754750 | |
| ≥97.0% (T) |
| 81998-04-1 | |
| White-Yellow | |
| MFCD02916382 | |
| 4-Hydroxymethyl-4′C-methyl-2,2′C-bipyridine, 4′C-Methyl-2,2′C-bipyridine-4-methanol | |
| CC1=CC(=NC=C1)C1=NC=CC(CO)=C1 | |
| 200.24 | |
| 200.24 | |
| Crystalline Powder |
Chemical Identifiers
| 81998-04-1 | |
| 200.24 | |
| GJCOKGHYIMLMPB-UHFFFAOYSA-N | |
| 2754750 | |
| CC1=CC(=NC=C1)C1=NC=CC(CO)=C1 |
| C12H12N2O | |
| MFCD02916382 | |
| 4-Hydroxymethyl-4′C-methyl-2,2′C-bipyridine, 4′C-Methyl-2,2′C-bipyridine-4-methanol | |
| {4'-methyl-[2,2'-bipyridin]-4-yl}methanol |
Safety and Handling
TSCA : No