missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxymethyl-4'-methyl-2,2'-bipyridyl 97.0+%, TCI America™

$231.02 - $783.54

Chemical Identifiers

CAS 81998-04-1
Molecular Formula C12H12N2O
Molecular Weight (g/mol) 200.24
MDL Number MFCD02916382
InChI Key GJCOKGHYIMLMPB-UHFFFAOYSA-N
Synonym 4-Hydroxymethyl-4′C-methyl-2,2′C-bipyridine, 4′C-Methyl-2,2′C-bipyridine-4-methanol
PubChem CID 2754750
IUPAC Name {4'-methyl-[2,2'-bipyridin]-4-yl}methanol
SMILES CC1=CC(=NC=C1)C1=NC=CC(CO)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H1347200MG
View Documents
TCI America
H1347200MG
200 mg
Each for $231.02
Only null left
 
H13471G
View Documents
TCI America
H13471G
1 g
Each for $783.54
Only null left
 
Description

Description

Specifications

Chemical Identifiers

81998-04-1
200.24
GJCOKGHYIMLMPB-UHFFFAOYSA-N
2754750
CC1=CC(=NC=C1)C1=NC=CC(CO)=C1
C12H12N2O
MFCD02916382
4-Hydroxymethyl-4′C-methyl-2,2′C-bipyridine, 4′C-Methyl-2,2′C-bipyridine-4-methanol
{4'-methyl-[2,2'-bipyridin]-4-yl}methanol

Specifications

81998-04-1
White-Yellow
MFCD02916382
4-Hydroxymethyl-4′C-methyl-2,2′C-bipyridine, 4′C-Methyl-2,2′C-bipyridine-4-methanol
CC1=CC(=NC=C1)C1=NC=CC(CO)=C1
200.24
200.24
Crystalline Powder
121°C
C12H12N2O
200 mg
GJCOKGHYIMLMPB-UHFFFAOYSA-N
{4'-methyl-[2,2'-bipyridin]-4-yl}methanol
2754750
≥97.0% (T)
4-Hydroxymethyl-4′-methyl-2,2′-bipyridyl
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'