missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxybenzotrifluoride 98.0+%, TCI America™
Supplier: TCI America H06445G
Specifications
| 4-Hydroxybenzotrifluoride | |
| 46°C | |
| C7H5F3O | |
| 5 g | |
| BAYGVMXZJBFEMB-UHFFFAOYSA-N | |
| 4-(trifluoromethyl)phenol | |
| 67874 | |
| 162.11 | |
| Crystalline Lumps |
| 402-45-9 | |
| 78°C | |
| MFCD00002363 | |
| 4-trifluoromethyl phenol, 4-hydroxybenzotrifluoride, alpha,alpha,alpha-trifluoro-p-cresol, p-trifluoromethylphenol, p-hydroxybenzotrifluoride, phenol, 4-trifluoromethyl, a,a,a-trifluoro-p-cresol, 4-trifluoromethyl-phenol, para-trifluoromethylphenol | |
| C1=CC(=CC=C1C(F)(F)F)O | |
| 162.111 | |
| CHEBI:42578 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 402-45-9 | |
| 162.111 | |
| BAYGVMXZJBFEMB-UHFFFAOYSA-N | |
| 67874 | |
| 4-(trifluoromethyl)phenol |
| C7H5F3O | |
| MFCD00002363 | |
| 4-trifluoromethyl phenol, 4-hydroxybenzotrifluoride, alpha,alpha,alpha-trifluoro-p-cresol, p-trifluoromethylphenol, p-hydroxybenzotrifluoride, phenol, 4-trifluoromethyl, a,a,a-trifluoro-p-cresol, 4-trifluoromethyl-phenol, para-trifluoromethylphenol | |
| CHEBI:42578 | |
| C1=CC(=CC=C1C(F)(F)F)O |
Safety and Handling
EINECSNumber : (9)-0915
TSCA : No
Recommended Storage : Refrigerator