missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzotrifluoride 98.0+%, TCI America™

Catalog No. p-7130655
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H06445G 5 g
H064425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H06445G Supplier TCI America Supplier No. H06445G
Only null left

Chemical Identifiers

CAS 402-45-9
Molecular Formula C7H5F3O
Molecular Weight (g/mol) 162.111
MDL Number MFCD00002363
InChI Key BAYGVMXZJBFEMB-UHFFFAOYSA-N
Synonym 4-trifluoromethyl phenol, 4-hydroxybenzotrifluoride, alpha,alpha,alpha-trifluoro-p-cresol, p-trifluoromethylphenol, p-hydroxybenzotrifluoride, phenol, 4-trifluoromethyl, a,a,a-trifluoro-p-cresol, 4-trifluoromethyl-phenol, para-trifluoromethylphenol
PubChem CID 67874
ChEBI CHEBI:42578
IUPAC Name 4-(trifluoromethyl)phenol
SMILES C1=CC(=CC=C1C(F)(F)F)O

Specifications

Melting Point 46°C
Boiling Point 78°C
Quantity 5 g
Formula Weight 162.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Hydroxybenzotrifluoride
EINECSNumber (9)-0915
TSCA No
Recommended Storage Refrigerator