missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzophenone 98.0+%, TCI America™

Catalog No. p-7130420
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H022225G 25 g
H0222500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H022225G Supplier TCI America Supplier No. H022225G
Only null left

Chemical Identifiers

CAS 1137-42-4
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.22
MDL Number MFCD00002355
InChI Key NPFYZDNDJHZQKY-UHFFFAOYSA-N
Synonym 4-hydroxybenzophenone, 4-benzoylphenol, 4-hydroxyphenyl phenyl methanone, p-benzoylphenol, p-hydroxybenzophenone, methanone, 4-hydroxyphenyl phenyl, 4-hydroxyphenyl phenylmethanone, benzophenone, 4-hydroxy, unii-04r2lws0ms, 4'-hydroxybenzophenone
PubChem CID 14347
ChEBI CHEBI:34421
IUPAC Name 4-benzoylphenol
SMILES OC1=CC=C(C=C1)C(=O)C1=CC=CC=C1

Specifications

Melting Point 134°C
Quantity 25 g
Formula Weight 198.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxybenzophenone
RTECSNumber PC4959775
TSCA Yes