missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzoic Acid 99.0+%, TCI America™

Catalog No. p-7130406
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Catalog No. Quantity
H020725G 25 g
H0207100G 100 g
H0207500G 500 g
3 options

Catalog No. H020725G

Supplier: TCI America H020725G

Only null left

Chemical Identifiers

CAS 99-96-7
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00002547
InChI Key FJKROLUGYXJWQN-UHFFFAOYSA-N
Synonym p-hydroxybenzoic acid, 4-carboxyphenol, benzoic acid, 4-hydroxy, p-salicylic acid, benzoic acid, p-hydroxy, para-hydroxybenzoic acid, 4-hydroxybenzoesaeure, p-carboxyphenol, 4-hydroxy benzoic acid, p-oxybenzoesaure german
PubChem CID 135
ChEBI CHEBI:30763
IUPAC Name 4-hydroxybenzoic acid
SMILES OC(=O)C1=CC=C(O)C=C1

Specifications

Melting Point 215°C
Color White
Quantity 25 g
Formula Weight 138.12
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxybenzoic Acid
EINECSNumber (3)-1640
RTECSNumber DH1925000
TSCA Yes