accessibility menu, dialog, popup

UserName

HOOOH

CAS RN 99-96-7

IUPAC Name: 4-hydroxybenzoic acid
Synonyms: P-hydroxybenzoic acid p-salicylic acid
Molecular Weight (g/mol): 138.12
Molecular Formula: C7H6O3
InChi Key: FJKROLUGYXJWQN-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(O)C=C1
Percent Purity 
  • (3)
  • (7)
  • (3)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (6)
  • (4)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (4)
  • (4)
  • (3)
  • (6)
  • (3)
  • (7)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (6)
  • (4)
  • (4)