missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzeneboronic acid pinacol ester, 97%

Catalog No. AAH2783603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2783603 1 g
AAH2783606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2783603 Supplier Thermo Scientific Chemicals Supplier No. H2783603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 269409-70-3 | C12H17BO3 | 220.075 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 269409-70-3
Molecular Formula C12H17BO3
Molecular Weight (g/mol) 220.075
MDL Number MFCD02093756
InChI Key BICZJRAGTCRORZ-UHFFFAOYSA-N
Synonym 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-hydroxyphenylboronic acid pinacol ester, 4-hydroxyphenylboronic acid, pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-hydroxyphenylboronic acid pinacolate, 2-4-hydroxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-hydroxyphenylboronicacidpinacolester, 4-hydroxybenzeneboronic acid pinacol ester, phenol, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-hydroxyphenylboronic acid pinacol cyclic ester
PubChem CID 2734624
IUPAC Name 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)O

Specifications

Melting Point 114°C to 117°C
Quantity 1 g
Formula Weight 220.07
Percent Purity 97%
Chemical Name or Material 4-Hydroxybenzeneboronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only