CAS RN 269409-70-3
4-Hydroxybenzeneboronic acid pinacol ester, 97%
CAS: 269409-70-3 Molecular Formula: C12H17BO3 Molecular Weight (g/mol): 220.075 MDL Number: MFCD02093756 InChI Key: BICZJRAGTCRORZ-UHFFFAOYSA-N Synonym: 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol,4-hydroxyphenylboronic acid pinacol ester,4-hydroxyphenylboronic acid, pinacol ester,4-tetramethyl-1,3,2-dioxaborolan-2-yl phenol,4-hydroxyphenylboronic acid pinacolate,2-4-hydroxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,4-hydroxyphenylboronicacidpinacolester,4-hydroxybenzeneboronic acid pinacol ester,phenol, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl,4-hydroxyphenylboronic acid pinacol cyclic ester PubChem CID: 2734624 IUPAC Name: 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol SMILES: B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)O
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol, 97%
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol 98.0+%, TCI America™
CAS: 269409-70-3 Molecular Formula: C12H17BO3 Molecular Weight (g/mol): 220.075 MDL Number: MFCD02093756 InChI Key: BICZJRAGTCRORZ-UHFFFAOYSA-N Synonym: 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol,4-hydroxyphenylboronic acid pinacol ester,4-hydroxyphenylboronic acid, pinacol ester,4-tetramethyl-1,3,2-dioxaborolan-2-yl phenol,4-hydroxyphenylboronic acid pinacolate,2-4-hydroxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,4-hydroxyphenylboronicacidpinacolester,4-hydroxybenzeneboronic acid pinacol ester,phenol, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl,4-hydroxyphenylboronic acid pinacol cyclic ester PubChem CID: 2734624 IUPAC Name: 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol SMILES: B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)O