missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals (-)-4-Hydroxy-D-phenylglycine, 98+%

CAS: 22818-40-2 | C8H9NO3 | 167.164 g/mol

Supplier:  Thermo Scientific Chemicals L0719014

 View more versions of this product

Catalog No. AAL0719014


Only null left

Description

Description

(-)-4-Hydroxy-D-phenylglycine is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(-)-4-Hydroxy-D-phenylglycine is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water (5 g/L at 20°C) and 1 mol/l HCl.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

∽240°C (decomposition)
22818-40-2
MFCD00004262
d-4-hydroxyphenylglycine, 4-hydroxy-d-phenylglycine, d--4-hydroxyphenylglycine, h-d-phg 4-oh-oh, d-n-4-hydroxyphenyl glycine, 2r-2-amino-2-4-hydroxyphenyl acetic acid, r-2-amino-2-4-hydroxyphenyl acetic acid, r-alpha-amino-4-hydroxybenzeneacetic acid, d---4-hydroxyphenylglycine, unii-pcm9oix717
LJCWONGJFPCTTL-SSDOTTSWSA-N
(2R)-2-amino-2-(4-hydroxyphenyl)acetic acid
167.164
CHEBI:15695
167.16
(-)-4-Hydroxy-D-phenylglycine
C8H9NO3
2210998
Soluble in water (5g/L at 20°C) and 1 mol/l HCl.
C1=CC(=CC=C1C(C(=O)O)N)O
25 g
89853
−156° (c=1 in 1N HCl)
≥98%

Chemical Identifiers

22818-40-2
167.164
LJCWONGJFPCTTL-SSDOTTSWSA-N
89853
(2R)-2-amino-2-(4-hydroxyphenyl)acetic acid
C8H9NO3
MFCD00004262
d-4-hydroxyphenylglycine, 4-hydroxy-d-phenylglycine, d--4-hydroxyphenylglycine, h-d-phg 4-oh-oh, d-n-4-hydroxyphenyl glycine, 2r-2-amino-2-4-hydroxyphenyl acetic acid, r-2-amino-2-4-hydroxyphenyl acetic acid, r-alpha-amino-4-hydroxybenzeneacetic acid, d---4-hydroxyphenylglycine, unii-pcm9oix717
CHEBI:15695
C1=CC(=CC=C1C(C(=O)O)N)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 245-247-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only