accessibility menu, dialog, popup

UserName

NH2(R)OHOOH

CAS RN 22818-40-2

IUPAC Name: (2R)-2-amino-2-(4-hydroxyphenyl)acetic acid
Synonyms: D-4-hydroxyphenylglycine D-N-(4-hydroxyphenyl)glycine
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: LJCWONGJFPCTTL-SSDOTTSWSA-N
SMILES: N[C@@H](C(O)=O)C1=CC=C(O)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (4)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)