missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals (-)-4-Hydroxy-D-phenylglycine, 98+%

Catalog No. p-7044575
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0719006 5 g
AAL0719014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0719006 Supplier Thermo Scientific Chemicals Supplier No. L0719006
Only null left

CAS: 22818-40-2 | C8H9NO3 | 167.164 g/mol

(-)-4-Hydroxy-D-phenylglycine is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(-)-4-Hydroxy-D-phenylglycine is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water (5 g/L at 20°C) and 1 mol/l HCl.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 22818-40-2
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00004262
InChI Key LJCWONGJFPCTTL-SSDOTTSWSA-N
Synonym d-4-hydroxyphenylglycine, 4-hydroxy-d-phenylglycine, d--4-hydroxyphenylglycine, h-d-phg 4-oh-oh, d-n-4-hydroxyphenyl glycine, 2r-2-amino-2-4-hydroxyphenyl acetic acid, r-2-amino-2-4-hydroxyphenyl acetic acid, r-alpha-amino-4-hydroxybenzeneacetic acid, d---4-hydroxyphenylglycine, unii-pcm9oix717
PubChem CID 89853
ChEBI CHEBI:15695
IUPAC Name (2R)-2-amino-2-(4-hydroxyphenyl)acetic acid
SMILES C1=CC(=CC=C1C(C(=O)O)N)O

Specifications

Melting Point ∽240°C (decomposition)
Beilstein 2210998
Solubility Information Soluble in water (5g/L at 20°C) and 1 mol/l HCl.
Quantity 5 g
Formula Weight 167.16
Optical Rotation −156° (c=1 in 1N HCl)
Percent Purity ≥98%
Chemical Name or Material (-)-4-Hydroxy-D-phenylglycine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 245-247-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only