missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxy-4'-nitrobiphenyl 98.0+%, TCI America™
Supplier: TCI America H07475G
Specifications
| 4-Hydroxy-4′-nitrobiphenyl | |
| 207°C | |
| C12H9NO3 | |
| 5 g | |
| ZNDJDQOECGBUNK-UHFFFAOYSA-N | |
| 4'-nitro-[1,1'-biphenyl]-4-ol | |
| 138078 | |
| ≥98.0% (GC) |
| 3916-44-7 | |
| Yellow | |
| MFCD00047742 | |
| 4′-Nitrobiphenyl-4-ol | |
| OC1=CC=C(C=C1)C1=CC=C(C=C1)[N+]([O-])=O | |
| 215.21 | |
| 215.21 | |
| Crystalline Powder |
Chemical Identifiers
| 3916-44-7 | |
| 215.21 | |
| ZNDJDQOECGBUNK-UHFFFAOYSA-N | |
| 4'-nitro-[1,1'-biphenyl]-4-ol |
| C12H9NO3 | |
| MFCD00047742 | |
| 138078 | |
| OC1=CC=C(C=C1)C1=CC=C(C=C1)[N+]([O-])=O |
Safety and Handling
TSCA : No