accessibility menu, dialog, popup

UserName

OHNOO

CAS RN 3916-44-7

IUPAC Name: 4'-nitro-[1,1'-biphenyl]-4-ol
Synonyms: 4'-nitro-[1,1'-biphenyl]-4-ol
Molecular Weight (g/mol): 215.21
Molecular Formula: C12H9NO3
InChi Key: ZNDJDQOECGBUNK-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results