missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxy-4'-nitrobiphenyl 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 207°C |
| Color | Yellow |
| Quantity | 1 g |
| Synonym | 4′C-Nitrobiphenyl-4-ol |
| Formula Weight | 215.21 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Hydroxy-4′-nitrobiphenyl |
Safety and Handling
| TSCA | No |