missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-nitrophenylacetic Acid 98.0+%, TCI America™

Catalog No. H13621G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13621G 1 g
H13625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. H13621G missing translation for 'mfr' TCI America missing translation for 'supplierNo' H13621G

Chemical Identifiers

CAS 10463-20-4
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.146
MDL Number MFCD00007122
InChI Key QBHBHOSRLDPIHG-UHFFFAOYSA-N
Synonym 4-hydroxy-3-nitrophenylacetic acid, 2-4-hydroxy-3-nitrophenyl acetic acid, 4-hydroxy-3-nitrophenyl acetic acid, 3-nitro-4-hydroxyphenylacetic acid, 3-nitro-4-hydroxy phenylacetic acid, benzeneacetic acid, 4-hydroxy-3-nitro, no2hpa, hydroxynitrophenylaceticacid, acmc-2098cj, epitope id:158629
PubChem CID 447364
ChEBI CHEBI:546274
SMILES C1=CC(=C(C=C1CC(=O)O)[N+](=O)[O-])O

Specifications

Melting Point 146°C
Color Yellow
Quantity 1 g
Formula Weight 197.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-3-nitrophenylacetic Acid
TSCA No
Recommended Storage Freezer