accessibility menu, dialog, popup

UserName

OOONOO

CAS RN 10463-20-4

IUPAC Name: 2-(3-nitro-4-oxidophenyl)acetate
Synonyms: 2-(3-nitro-4-oxidophenyl)acetate
Molecular Weight (g/mol): 195.13
Molecular Formula: C8H5NO5
InChi Key: QBHBHOSRLDPIHG-UHFFFAOYSA-L
SMILES: [O-]C(=O)CC1=CC=C([O-])C(=C1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)