missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-nitrophenylacetic acid, 99%

Catalog No. AAA1385806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1385806 5 g
AAA1385814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1385806 Supplier Thermo Scientific Chemicals Supplier No. A1385806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 10463-20-4 | C8H7NO5 | 197.146 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10463-20-4
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.146
MDL Number MFCD00007122
InChI Key QBHBHOSRLDPIHG-UHFFFAOYSA-N
Synonym 4-hydroxy-3-nitrophenylacetic acid, 2-4-hydroxy-3-nitrophenyl acetic acid, 4-hydroxy-3-nitrophenyl acetic acid, 3-nitro-4-hydroxyphenylacetic acid, 3-nitro-4-hydroxy phenylacetic acid, benzeneacetic acid, 4-hydroxy-3-nitro, no2hpa, hydroxynitrophenylaceticacid, acmc-2098cj, epitope id:158629
PubChem CID 447364
ChEBI CHEBI:546274
SMILES C1=CC(=C(C=C1CC(=O)O)[N+](=O)[O-])O

Specifications

Melting Point 146°C to 148°C
Quantity 5 g
Beilstein 2696044
Formula Weight 197.15
Percent Purity 99%
Chemical Name or Material 4-Hydroxy-3-nitrophenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 233-948-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only