missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-nitrobenzaldehyde, 97%

CAS: 3011-34-5 | C7H5NO4 | 167.12 g/mol

$137.74 - $137.74

Chemical Identifiers

CAS 3011-34-5
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00007117
InChI Key YTHJCZRFJGXPTL-UHFFFAOYSA-N
Synonym 4-hydroxy-3-nitrobenzaldehyde, benzaldehyde, 4-hydroxy-3-nitro, 3-nitro-4-hydroxybenzaldehyde, 4-formyl-2-nitrophenol, 4-hydroxy-3-nitro-benzaldehyde, pubchem8135, acmc-1cjnu, d03vzf, 4-hydroxy-3-nitrobenzaldeyde, ksc223k5f
PubChem CID 18169
SMILES C1=CC(=C(C=C1C=O)[N+](=O)[O-])O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC162890050
SDS View Documents
Thermo Scientific Chemicals
162890050
5 g Glass bottle
Each for $137.74
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3011-34-5
167.12
YTHJCZRFJGXPTL-UHFFFAOYSA-N
18169
C7H5NO4
MFCD00007117
4-hydroxy-3-nitrobenzaldehyde, benzaldehyde, 4-hydroxy-3-nitro, 3-nitro-4-hydroxybenzaldehyde, 4-formyl-2-nitrophenol, 4-hydroxy-3-nitro-benzaldehyde, pubchem8135, acmc-1cjnu, d03vzf, 4-hydroxy-3-nitrobenzaldeyde, ksc223k5f
C1=CC(=C(C=C1C=O)[N+](=O)[O-])O

Specifications

3011-34-5
100.0
Brown to Brown-Yellow
96% min. (GC)
C7H5NO4
MFCD00007117
08, 83
YTHJCZRFJGXPTL-UHFFFAOYSA-N
167.12
167.12
Powder
96.0
139°C to 144°C
Authentic
Glass bottle
HOC6H3(NO2)CHO
5 g
4-hydroxy-3-nitrobenzaldehyde, benzaldehyde, 4-hydroxy-3-nitro, 3-nitro-4-hydroxybenzaldehyde, 4-formyl-2-nitrophenol, 4-hydroxy-3-nitro-benzaldehyde, pubchem8135, acmc-1cjnu, d03vzf, 4-hydroxy-3-nitrobenzaldeyde, ksc223k5f
C1=CC(=C(C=C1C=O)[N+](=O)[O-])O
18169
97%
4-Hydroxy-3-nitrobenzaldehyde, 97%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 221-141-6

SDS
Documents

Documents

RUO – Research Use Only