missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-nitrobenzaldehyde, 97%

Catalog No. AC162890050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC162890050 5 g Glass bottle
AC162890250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC162890050 Supplier Thermo Scientific Chemicals Supplier No. 162890050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3011-34-5 | C7H5NO4 | 167.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3011-34-5
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00007117
InChI Key YTHJCZRFJGXPTL-UHFFFAOYSA-N
Synonym 4-hydroxy-3-nitrobenzaldehyde, benzaldehyde, 4-hydroxy-3-nitro, 3-nitro-4-hydroxybenzaldehyde, 4-formyl-2-nitrophenol, 4-hydroxy-3-nitro-benzaldehyde, pubchem8135, acmc-1cjnu, d03vzf, 4-hydroxy-3-nitrobenzaldeyde, ksc223k5f
PubChem CID 18169
SMILES C1=CC(=C(C=C1C=O)[N+](=O)[O-])O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 139°C to 144°C
Color Brown to Brown-Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula HOC6H3(NO2)CHO
Quantity 5 g
Beilstein 08, 83
Formula Weight 167.12
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Hydroxy-3-nitrobenzaldehyde, 97%
Show More Show Less
EINECSNumber 221-141-6

RUO – Research Use Only