missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-methoxyphenylpyruvic Acid 95.0+%, TCI America™

Catalog No. H0539100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0539100MG 100 mg
H05391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0539100MG Supplier TCI America Supplier No. H0539100MG

Chemical Identifiers

CAS 1081-71-6
Molecular Formula C10H10O5
Molecular Weight (g/mol) 210.185
MDL Number MFCD00056707
InChI Key YGQHQTMRZPHIBB-UHFFFAOYSA-N
PubChem CID 14124
IUPAC Name 3-(4-hydroxy-3-methoxyphenyl)-2-oxopropanoic acid
SMILES COC1=C(C=CC(=C1)CC(=O)C(=O)O)O

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 210.19
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-3-methoxyphenylpyruvic Acid