Learn More
4-Hydroxy-3,5-diiodobenzonitrile, 97%
CAS: 1689-83-4 | C7H3I2NO | 370.92 g/mol
$476.63 - $476.63
Chemical Identifiers
| CAS | 1689-83-4 |
|---|---|
| Molecular Formula | C7H3I2NO |
| Molecular Weight (g/mol) | 370.92 |
| MDL Number | MFCD00016422 |
| InChI Key | NRXQIUSYPAHGNM-UHFFFAOYSA-N |
| Synonym | ioxynil, actril, bentrol, 3,5-diiodo-4-hydroxybenzonitrile, cipotril, trevespan, certrol, joxynil, loxynil, iotox |
| PubChem CID | 15530 |
| ChEBI | CHEBI:81821 |
| IUPAC Name | 4-hydroxy-3,5-diiodobenzonitrile |
| SMILES | OC1=C(I)C=C(C=C1I)C#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1392914
|
Thermo Scientific Chemicals
A1392914 |
25 g |
Each for $476.63
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1689-83-4 | |
| 370.92 | |
| NRXQIUSYPAHGNM-UHFFFAOYSA-N | |
| 15530 | |
| 4-hydroxy-3,5-diiodobenzonitrile |
| C7H3I2NO | |
| MFCD00016422 | |
| ioxynil, actril, bentrol, 3,5-diiodo-4-hydroxybenzonitrile, cipotril, trevespan, certrol, joxynil, loxynil, iotox | |
| CHEBI:81821 | |
| OC1=C(I)C=C(C=C1I)C#N |
Specifications
| 1689-83-4 | |
| Odorless | |
| MFCD00016422 | |
| UN3439 | |
| Light sensitive | |
| NRXQIUSYPAHGNM-UHFFFAOYSA-N | |
| 4-hydroxy-3,5-diiodobenzonitrile | |
| 15530 | |
| 370.92 | |
| 4-Hydroxy-3,5-diiodobenzonitrile |
| 200°C to 207°C | |
| C7H3I2NO | |
| 25 g | |
| 2364041 | |
| ioxynil, actril, bentrol, 3,5-diiodo-4-hydroxybenzonitrile, cipotril, trevespan, certrol, joxynil, loxynil, iotox | |
| OC1=C(I)C=C(C=C1I)C#N | |
| 370.92 | |
| CHEBI:81821 | |
| 97% |
Safety and Handling
GHS H Statement
H301-H331-H361-H373-H312-H319
Toxic if swallowed.
Toxic if inhaled.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes serious eye irritation.
P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P312-P330-P363-P501c
H301+H331-H312-H319-H361d-H373-H500
DOTInformation : Hazard Class: 6.1; Packaging Group: II
EINECSNumber : 216-881-1
RTECSNumber : DI4025000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only