CAS RN 1689-83-4
IUPAC Name:
4-hydroxy-3,5-diiodobenzonitrile
Synonyms:
ioxynil
Molecular Weight (g/mol):
370.92
Molecular Formula:
C7H3I2NO
InChi Key:
NRXQIUSYPAHGNM-UHFFFAOYSA-N
SMILES:
OC1=C(I)C=C(C=C1I)C#N
1
–
1
of
1
results
1
Promotions Available
4-Hydroxy-3,5-diiodobenzonitrile, 97%
4-Hydroxy-3,5-diiodobenzonitrile, 97%
CAS: 1689-83-4 Molecular Formula: C7H3I2NO Molecular Weight (g/mol): 370.92 MDL Number: MFCD00016422 InChI Key: NRXQIUSYPAHGNM-UHFFFAOYSA-N Synonym: ioxynil,actril,bentrol,3,5-diiodo-4-hydroxybenzonitrile,cipotril,trevespan,certrol,joxynil,loxynil,iotox PubChem CID: 15530 ChEBI: CHEBI:81821 IUPAC Name: 4-hydroxy-3,5-diiodobenzonitrile SMILES: OC1=C(I)C=C(C=C1I)C#N