missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2-(trifluoromethyl)benzonitrile, 98%

CAS: 320-42-3 | C8H4F3NO | 187.121 g/mol

$176.18 - $211.84

Chemical Identifiers

CAS 320-42-3
Molecular Formula C8H4F3NO
Molecular Weight (g/mol) 187.121
MDL Number MFCD06797340
InChI Key MDBVIOUGHSKRMT-UHFFFAOYSA-N
Synonym 4-hydroxy-2-trifluoromethyl benzonitrile, 4-hydroxy-2-trifluoroemthyl benzonitrile, pubchem4807, 2-cyano-5-hydroxybenzotrifluoride, 4-cyano-3-trifluoromethyl phenol, 4-hydroxy-2-trifluoromethyl-benzonitrile, benzonitrile, 4-hydroxy-2-trifluoromethyl, 4-hydroxy-2-trifluoromethyl benzenecarbonitrile
PubChem CID 2783153
IUPAC Name 4-hydroxy-2-(trifluoromethyl)benzonitrile
SMILES C1=CC(=C(C=C1O)C(F)(F)F)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH32158MD
View Documents
Thermo Scientific Chemicals
H32158MD
250 mg
Each for $176.18
Only null left
 
AAH3215803
View Documents
Thermo Scientific Chemicals
H3215803
1 g
Each for $211.84
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

320-42-3
187.121
MDBVIOUGHSKRMT-UHFFFAOYSA-N
2783153
C1=CC(=C(C=C1O)C(F)(F)F)C#N
C8H4F3NO
MFCD06797340
4-hydroxy-2-trifluoromethyl benzonitrile, 4-hydroxy-2-trifluoroemthyl benzonitrile, pubchem4807, 2-cyano-5-hydroxybenzotrifluoride, 4-cyano-3-trifluoromethyl phenol, 4-hydroxy-2-trifluoromethyl-benzonitrile, benzonitrile, 4-hydroxy-2-trifluoromethyl, 4-hydroxy-2-trifluoromethyl benzenecarbonitrile
4-hydroxy-2-(trifluoromethyl)benzonitrile

Specifications

320-42-3
C8H4F3NO
250 mg
MDBVIOUGHSKRMT-UHFFFAOYSA-N
4-hydroxy-2-(trifluoromethyl)benzonitrile
2783153
98%
120°C to 124°C
MFCD06797340
4-hydroxy-2-trifluoromethyl benzonitrile, 4-hydroxy-2-trifluoroemthyl benzonitrile, pubchem4807, 2-cyano-5-hydroxybenzotrifluoride, 4-cyano-3-trifluoromethyl phenol, 4-hydroxy-2-trifluoromethyl-benzonitrile, benzonitrile, 4-hydroxy-2-trifluoromethyl, 4-hydroxy-2-trifluoromethyl benzenecarbonitrile
C1=CC(=C(C=C1O)C(F)(F)F)C#N
187.121
187.12
4-Hydroxy-2-(trifluoromethyl)benzonitrile
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only