missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2-(trifluoromethyl)benzonitrile, 98%

CAS: 320-42-3 | C8H4F3NO | 187.121 g/mol

Supplier:  Thermo Scientific Chemicals H3215803

 View more versions of this product

Catalog No. AAH3215803


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Hydroxy-2-(trifluoromethyl)benzonitrile
120°C to 124°C
MFCD06797340
4-hydroxy-2-trifluoromethyl benzonitrile, 4-hydroxy-2-trifluoroemthyl benzonitrile, pubchem4807, 2-cyano-5-hydroxybenzotrifluoride, 4-cyano-3-trifluoromethyl phenol, 4-hydroxy-2-trifluoromethyl-benzonitrile, benzonitrile, 4-hydroxy-2-trifluoromethyl, 4-hydroxy-2-trifluoromethyl benzenecarbonitrile
C1=CC(=C(C=C1O)C(F)(F)F)C#N
187.121
187.12
320-42-3
C8H4F3NO
1 g
MDBVIOUGHSKRMT-UHFFFAOYSA-N
4-hydroxy-2-(trifluoromethyl)benzonitrile
2783153
98%

Chemical Identifiers

320-42-3
187.121
MDBVIOUGHSKRMT-UHFFFAOYSA-N
2783153
C1=CC(=C(C=C1O)C(F)(F)F)C#N
C8H4F3NO
MFCD06797340
4-hydroxy-2-trifluoromethyl benzonitrile, 4-hydroxy-2-trifluoroemthyl benzonitrile, pubchem4807, 2-cyano-5-hydroxybenzotrifluoride, 4-cyano-3-trifluoromethyl phenol, 4-hydroxy-2-trifluoromethyl-benzonitrile, benzonitrile, 4-hydroxy-2-trifluoromethyl, 4-hydroxy-2-trifluoromethyl benzenecarbonitrile
4-hydroxy-2-(trifluoromethyl)benzonitrile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only