missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2-methylbenzoic Acid 98.0+%, TCI America™

Catalog No. H13115G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13115G 5 g
H131125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H13115G Supplier TCI America Supplier No. H13115G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 578-39-2
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD02182261
InChI Key BBMFSGOFUHEVNP-UHFFFAOYSA-N
Synonym 4-hydroxy-o-toluic acid, 4,2-cresotic acid, 2-methyl-4-hydroxybenzoic acid, 4-hydroxy-2-methyl-benzoic acid, benzoic acid, 4-hydroxy-2-methyl, 2-methyl-4-hydroxybenzoicacid, acmc-209m0q, methyl p-hydroxybenzoic acid, ksc494s9j, 4-hydroxy-2-methylbenzoicacid
PubChem CID 68475
IUPAC Name 4-hydroxy-2-methylbenzoic acid
SMILES CC1=C(C=CC(=C1)O)C(=O)O

Specifications

Melting Point 179°C
Quantity 5 g
Formula Weight 152.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-2-methylbenzoic Acid