accessibility menu, dialog, popup

UserName

CAS RN 578-39-2

IUPAC Name: 4-hydroxy-2-methylbenzoic acid
Synonyms: 4-hydroxy-2-methylbenzoic acid
Molecular Weight (g/mol): 152.15
Molecular Formula: C8H8O3
InChi Key: BBMFSGOFUHEVNP-UHFFFAOYSA-N
SMILES: CC1=CC(O)=CC=C1C(O)=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)