missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2-methylbenzoic acid, 98%, Thermo Scientific Chemicals

Catalog No. p-7040909
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6081009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6081009 Supplier Thermo Scientific Chemicals Supplier No. H6081009

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 578-39-2 | Molecular Formula: C8H8O3 | Molecular Weight: 152.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 578-39-2
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD02182261
InChI Key BBMFSGOFUHEVNP-UHFFFAOYSA-N
Synonym 4-hydroxy-o-toluic acid, 4,2-cresotic acid, 2-methyl-4-hydroxybenzoic acid, 4-hydroxy-2-methyl-benzoic acid, benzoic acid, 4-hydroxy-2-methyl, 2-methyl-4-hydroxybenzoicacid, acmc-209m0q, methyl p-hydroxybenzoic acid, ksc494s9j, 4-hydroxy-2-methylbenzoicacid
PubChem CID 68475
IUPAC Name 4-hydroxy-2-methylbenzoic acid
SMILES CC1=C(C=CC(=C1)O)C(=O)O

Specifications

Melting Point 177°C to 181°C
Quantity 10 g
Formula Weight 152.15
Percent Purity 98%
Chemical Name or Material 4-Hydroxy-2-methylbenzoic acid
Hazard Category H302-H318
Hazard Statement GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
EINECSNumber 209-422-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only