missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydrazinobenzenesulfonic Acid 98.0+%, TCI America™
Supplier: TCI America H1662100G
Specifications
| 4-Hydrazinobenzenesulfonic Acid | |
| 286°C | |
| C6H8N2O3S | |
| 100 g | |
| IOMZCWUHFGMSEJ-UHFFFAOYSA-N | |
| 4-hydrazinylbenzene-1-sulfonic acid | |
| 66825 | |
| ≥98.0% (HPLC,T) |
| 98-71-5 | |
| White-Yellow | |
| MFCD00025097 | |
| 4-hydrazinobenzenesulfonic acid, phenylhydrazine-4-sulfonic acid, benzenesulfonic acid, 4-hydrazino, p-hydrazinobenzenesulfonic acid, p-sulfophenylhydrazine, 4-sulfophenylhydrazine, 4-hydrazinobenzenesulphonic acid, phenylhydrazinesulfonic acid, phenylhydrazine-p-sulfonic acid, sulfanilic acid, n-amino | |
| NNC1=CC=C(C=C1)S(O)(=O)=O | |
| 188.20 | |
| 188.20 | |
| Crystalline Powder |
Chemical Identifiers
| 98-71-5 | |
| 188.20 | |
| IOMZCWUHFGMSEJ-UHFFFAOYSA-N | |
| 66825 | |
| NNC1=CC=C(C=C1)S(O)(=O)=O |
| C6H8N2O3S | |
| MFCD00025097 | |
| 4-hydrazinobenzenesulfonic acid, phenylhydrazine-4-sulfonic acid, benzenesulfonic acid, 4-hydrazino, p-hydrazinobenzenesulfonic acid, p-sulfophenylhydrazine, 4-sulfophenylhydrazine, 4-hydrazinobenzenesulphonic acid, phenylhydrazinesulfonic acid, phenylhydrazine-p-sulfonic acid, sulfanilic acid, n-amino | |
| 4-hydrazinylbenzene-1-sulfonic acid |
Safety and Handling
EINECSNumber : (3)-1997
RTECSNumber : DB6900000
TSCA : Yes