accessibility menu, dialog, popup

UserName

CAS RN 98-71-5

IUPAC Name: 4-hydrazinylbenzene-1-sulfonic acid
Synonyms: 4-hydrazinylbenzenesulfonic acid
Molecular Weight (g/mol): 188.2
Molecular Formula: C6H8N2O3S
InChi Key: IOMZCWUHFGMSEJ-UHFFFAOYSA-N
SMILES: NNC1=CC=C(C=C1)S(O)(=O)=O
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)