missing translation for 'onlineSavingsMsg'
Learn More

4-Hydrazinobenzenesulfonic acid hydrate, 98%

Catalog No. aaa1336222
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1336222 100 g
AAA1336206 5 g
AAA1336214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1336222 Supplier Thermo Scientific Chemicals Supplier No. A1336222
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 98-71-5 | C6H8N2O3S | 188.20 g/mol

4-Hydrazinobenzenesulfonic Acid is used as an intermediate to manufacture pharmaceuticals, dyes and other organic synthesis. They have active applications in organic synthesis for agrochemicals, pharmaceuticals, photographic, heat stabilizers, polymerization catalysts, flame-retardants, blowing agents for plastics, explosives, and dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydrazinobenzenesulfonic Acid is used as an intermediate to manufacture pharmaceuticals, dyes and other organic synthesis. They have active applications in organic synthesis for agrochemicals, pharmaceuticals, photographic, heat stabilizers, polymerization catalysts, flame-retardants, blowing agents for plastics, explosives, and dyes.

Solubility
It is soluble in water.

Notes
Stable under normal conditions.Keep away from strong oxidizing agents. Store in closed vessels.Hygroscopic

Chemical Identifiers

CAS 98-71-5
Molecular Formula C6H8N2O3S
Molecular Weight (g/mol) 188.20
MDL Number MFCD00025097
InChI Key IOMZCWUHFGMSEJ-UHFFFAOYSA-N
Synonym 4-hydrazinobenzenesulfonic acid, phenylhydrazine-4-sulfonic acid, benzenesulfonic acid, 4-hydrazino, p-hydrazinobenzenesulfonic acid, p-sulfophenylhydrazine, 4-sulfophenylhydrazine, 4-hydrazinobenzenesulphonic acid, phenylhydrazinesulfonic acid, phenylhydrazine-p-sulfonic acid, sulfanilic acid, n-amino
PubChem CID 66825
IUPAC Name 4-hydrazinylbenzenesulfonic acid
SMILES NNC1=CC=C(C=C1)S(O)(=O)=O

Specifications

Melting Point ∼286°C (decomposition)
Quantity 100 g
Beilstein 514779
Sensitivity Hygroscopic
Merck Index 14,4773
Solubility Information It is soluble in water.
Formula Weight 188.20 (Anhydrous)
Percent Purity 98%
Chemical Name or Material 4-Hydrazinobenzenesulfonic acid hydrate
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 202-694-2
RTECSNumber DB6900000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only