Learn More
4-Hydrazinobenzenesulfonic acid hydrate, 98%
CAS: 98-71-5 | C6H8N2O3S | 188.20 g/mol
Supplier: Thermo Scientific Chemicals A1336214
Description
4-Hydrazinobenzenesulfonic Acid is used as an intermediate to manufacture pharmaceuticals, dyes and other organic synthesis. They have active applications in organic synthesis for agrochemicals, pharmaceuticals, photographic, heat stabilizers, polymerization catalysts, flame-retardants, blowing agents for plastics, explosives, and dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Hydrazinobenzenesulfonic Acid is used as an intermediate to manufacture pharmaceuticals, dyes and other organic synthesis. They have active applications in organic synthesis for agrochemicals, pharmaceuticals, photographic, heat stabilizers, polymerization catalysts, flame-retardants, blowing agents for plastics, explosives, and dyes.
Solubility
It is soluble in water.
Notes
Stable under normal conditions.Keep away from strong oxidizing agents. Store in closed vessels.Hygroscopic
Specifications
| 4-Hydrazinobenzenesulfonic acid hydrate | |
| ∼286°C (decomposition) | |
| MFCD00025097 | |
| 514779 | |
| 14,4773 | |
| It is soluble in water. | |
| NNC1=CC=C(C=C1)S(O)(=O)=O | |
| 188.20 | |
| 188.20 (Anhydrous) |
| 98-71-5 | |
| C6H8N2O3S | |
| 25 g | |
| Hygroscopic | |
| 4-hydrazinobenzenesulfonic acid, phenylhydrazine-4-sulfonic acid, benzenesulfonic acid, 4-hydrazino, p-hydrazinobenzenesulfonic acid, p-sulfophenylhydrazine, 4-sulfophenylhydrazine, 4-hydrazinobenzenesulphonic acid, phenylhydrazinesulfonic acid, phenylhydrazine-p-sulfonic acid, sulfanilic acid, n-amino | |
| IOMZCWUHFGMSEJ-UHFFFAOYSA-N | |
| 4-hydrazinylbenzenesulfonic acid | |
| 66825 | |
| 98% |
Chemical Identifiers
| 98-71-5 | |
| 188.20 | |
| IOMZCWUHFGMSEJ-UHFFFAOYSA-N | |
| 66825 | |
| NNC1=CC=C(C=C1)S(O)(=O)=O |
| C6H8N2O3S | |
| MFCD00025097 | |
| 4-hydrazinobenzenesulfonic acid, phenylhydrazine-4-sulfonic acid, benzenesulfonic acid, 4-hydrazino, p-hydrazinobenzenesulfonic acid, p-sulfophenylhydrazine, 4-sulfophenylhydrazine, 4-hydrazinobenzenesulphonic acid, phenylhydrazinesulfonic acid, phenylhydrazine-p-sulfonic acid, sulfanilic acid, n-amino | |
| 4-hydrazinylbenzenesulfonic acid |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 202-694-2
RTECSNumber : DB6900000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only