missing translation for 'onlineSavingsMsg'
Learn More

4-(Hexyloxy)benzoic Acid 98.0+%, TCI America™

Catalog No. H0247500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0247500G 500 g
H024725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0247500G Supplier TCI America Supplier No. H0247500G

Chemical Identifiers

CAS 1142-39-8
Molecular Formula C13H18O3
Molecular Weight (g/mol) 222.284
MDL Number MFCD00013991
InChI Key HBQUXMZZODHFMJ-UHFFFAOYSA-N
PubChem CID 70834
IUPAC Name 4-hexoxybenzoic acid
SMILES CCCCCCOC1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 155°C
Color White
Quantity 500 g
Formula Weight 222.28
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Hexyloxy)benzoic Acid