accessibility menu, dialog, popup

UserName

H3COOHO

CAS RN 1142-39-8

IUPAC Name: 4-(hexyloxy)benzoic acid
Synonyms: 4-(hexyloxy)benzoic acid
Molecular Weight (g/mol): 222.28
Molecular Formula: C13H18O3
InChi Key: HBQUXMZZODHFMJ-UHFFFAOYSA-N
SMILES: CCCCCCOC1=CC=C(C=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results