missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hexylaniline 98.0+%, TCI America™
Supplier: TCI America H089025G
Specifications
| 4-Hexylaniline | |
| Yellow | |
| C12H19N | |
| 25 g | |
| OVEMTTZEBOCJDV-UHFFFAOYSA-N | |
| 4-hexylaniline | |
| 36382 | |
| ≥98.0% (GC,T) |
| 33228-45-4 | |
| 285°C | |
| MFCD00007927 | |
| p-hexylaniline, 4-n-hexylaniline, benzenamine, 4-hexyl, aniline, p-hexyl, 4-hexylbenzenamine, p-n-hexylaniline, unii-u3c0879zgc, 4-hexylphenylamine, 4-n-hexylaniline;, acmc-209hzh | |
| CCCCCCC1=CC=C(C=C1)N | |
| 177.291 | |
| 177.29 | |
| Liquid |
Chemical Identifiers
| 33228-45-4 | |
| 177.291 | |
| OVEMTTZEBOCJDV-UHFFFAOYSA-N | |
| 36382 | |
| CCCCCCC1=CC=C(C=C1)N |
| C12H19N | |
| MFCD00007927 | |
| p-hexylaniline, 4-n-hexylaniline, benzenamine, 4-hexyl, aniline, p-hexyl, 4-hexylbenzenamine, p-n-hexylaniline, unii-u3c0879zgc, 4-hexylphenylamine, 4-n-hexylaniline;, acmc-209hzh | |
| 4-hexylaniline |
Safety and Handling
EINECSNumber : (3)-0206
RTECSNumber : BY2305000
TSCA : No
Recommended Storage : Refrigerator