missing translation for 'onlineSavingsMsg'
Learn More

4-Hexylaniline 98.0+%, TCI America™

Catalog No. p-7130786
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08905G 5 g
H089025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08905G Supplier TCI America Supplier No. H08905G

Chemical Identifiers

CAS 33228-45-4
Molecular Formula C12H19N
Molecular Weight (g/mol) 177.291
MDL Number MFCD00007927
InChI Key OVEMTTZEBOCJDV-UHFFFAOYSA-N
Synonym p-hexylaniline, 4-n-hexylaniline, benzenamine, 4-hexyl, aniline, p-hexyl, 4-hexylbenzenamine, p-n-hexylaniline, unii-u3c0879zgc, 4-hexylphenylamine, 4-n-hexylaniline;, acmc-209hzh
PubChem CID 36382
IUPAC Name 4-hexylaniline
SMILES CCCCCCC1=CC=C(C=C1)N

Specifications

Color Yellow
Boiling Point 285°C
Quantity 5 g
Formula Weight 177.29
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Hexylaniline
EINECSNumber (3)-0206
RTECSNumber BY2305000
TSCA No
Recommended Storage Refrigerator