accessibility menu, dialog, popup

UserName

H3CNH2

CAS RN 33228-45-4

IUPAC Name: 4-hexylaniline
Synonyms: aniline, (p-hexyl)-
Molecular Weight (g/mol): 177.29
Molecular Formula: C12H19N
InChi Key: OVEMTTZEBOCJDV-UHFFFAOYSA-N
SMILES: CCCCCCC1=CC=C(N)C=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)