missing translation for 'onlineSavingsMsg'
Learn More

4-Guanidinobenzoic Acid Methanesulfonate 98.0+%, TCI America™

Catalog No. p-7130209
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G02485G 5 g
G024825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G02485G Supplier TCI America Supplier No. G02485G

Chemical Identifiers

CAS 148720-07-4
Molecular Formula C9H13N3O5S
Molecular Weight (g/mol) 275.28
MDL Number MFCD00191442
InChI Key NGBBXMQQIAFCGF-UHFFFAOYSA-N
PubChem CID 44630314
IUPAC Name 4-[(diaminomethylidene)amino]benzoic acid; methanesulfonic acid
SMILES CS(O)(=O)=O.NC(N)=NC1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 228°C
Color White
Quantity 5 g
Formula Weight 275.28
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Guanidinobenzoic Acid Methanesulfonate