accessibility menu, dialog, popup

UserName

CH3SOOHOOHONNH2H2N

CAS RN 148720-07-4

IUPAC Name: 4-[(diaminomethylidene)amino]benzoic acid; methanesulfonic acid
Synonyms: 4-[(diaminomethylidene)amino]benzoic acid; methanesulfonic acid
Molecular Weight (g/mol): 275.28
Molecular Formula: C9H13N3O5S
InChi Key: NGBBXMQQIAFCGF-UHFFFAOYSA-N
SMILES: CS(O)(=O)=O.NC(N)=NC1=CC=C(C=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results