missing translation for 'onlineSavingsMsg'
Learn More

4-Guanidinobenzoic Acid Hydrochloride 97.0+%, TCI America™

Catalog No. p-7130203
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G02385G 5 g
G023825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G02385G Supplier TCI America Supplier No. G02385G

Chemical Identifiers

CAS 42823-46-1
Molecular Formula C8H10ClN3O2
Molecular Weight (g/mol) 215.637
MDL Number MFCD00040587
InChI Key YETFLAUJROGBMC-UHFFFAOYSA-N
Synonym 4-guanidinobenzoic acid hydrochloride, n-4-carboxyphenyl guanidine hydrochloride, 4-guanidinobenzoic acid hcl, 4-guanidino benzoic acid hydrochloride, 4-amidinoamino benzoic acid, chloride, pubchem14981, pubchem14993, acmc-1an3m, ksc494a0j, 4-guanidino-benzoic acid-hcl salt
PubChem CID 3084875
IUPAC Name 4-(diaminomethylideneamino)benzoic acid;hydrochloride
SMILES C1=CC(=CC=C1C(=O)O)N=C(N)N.Cl

Specifications

Melting Point 276°C
Color White
Quantity 5 g
Formula Weight 215.64
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Guanidinobenzoic Acid Hydrochloride