accessibility menu, dialog, popup

UserName

HClNH2H2NNOHO

CAS RN 42823-46-1

IUPAC Name: hydrogen 4-[(diaminomethylidene)amino]benzoic acid chloride
Synonyms: hydrogen 4-[(diaminomethylidene)amino]benzoic acid chloride
Molecular Weight (g/mol): 215.64
Molecular Formula: C8H10ClN3O2
InChi Key: YETFLAUJROGBMC-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC(N)=NC1=CC=C(C=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results