missing translation for 'onlineSavingsMsg'
Learn More

Labconco 4 ft. Nexus Horizontal Clean Bench

Nexus Horizontal Clean Benches are designed to protect non-hazardous applications by providing an ISO Class 4 clean airflow environment. The large, 5 in. touchscreen interface features Nexus OS™ software that simplifies operation, maintains records and provides user customizaton of the system.

$17,680.84 - $23,623.82

Specifications

Product Type Horizontal Clean Bench
Coating Epoxy
Filter Type HEPA
Color Optic White
Height (English) 50.1 in.
View More Specs

Products 32
Catalog Number Mfr. No. Material Voltage Includes Depth (English) Interior Price Quantity & Availability  
Catalog Number Mfr. No. Material Voltage Includes Depth (English) Interior Price Quantity & Availability  
17195051
View Documents Promotion Details
Labconco
LCN5340000
Epoxy-coated Steel 115 V Clean Bench Only 21 in. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
17194997
View Documents Promotion Details
Labconco
5340010
Epoxy-coated Steel 230 V Clean Bench Only 21 in. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
17195052
View Documents Promotion Details
Labconco
5340100
Stainless Steel 115 V Clean Bench Only 21 in. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
17195002
View Documents Promotion Details
Labconco
LCN5340110
Stainless Steel 230 V Clean Bench Only 21 in. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
17195085
View Documents Promotion Details
Labconco
5340200
Epoxy-coated Steel 115 V UV Lamp 21 in. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
17195101
View Documents Promotion Details
Labconco
5340210
Epoxy-coated Steel 230 V UV Lamp 21 in. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
17195054
View Documents Promotion Details
Labconco
5340300
Stainless Steel 115 V UV Lamp 21 in. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
17194980
View Documents Promotion Details
Labconco
5340310
Stainless Steel 230 V UV Lamp 21 in. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
17195010
View Documents Promotion Details
Labconco
LCN5340500
Epoxy-coated Steel 115 V Base Stand 21 in. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
17195011
View Documents Promotion Details
Labconco
5340510
Epoxy-coated Steel 230 V Base Stand 21 in. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
17194991
View Documents Promotion Details
Labconco
5340600
Stainless Steel 115 V Base Stand 21 in. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
17195028
View Documents Promotion Details
Labconco
5340610
Stainless Steel 230 V Base Stand 21 in. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
17194998
View Documents Promotion Details
Labconco
5340700
Epoxy-coated Steel 115 V UV Lamp and Base Stand 21 in. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
17194964
View Documents Promotion Details
Labconco
5340710
Epoxy-coated Steel 230 V UV Lamp and Base Stand 21 in. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
17194981
View Documents Promotion Details
Labconco
5340800
Stainless Steel 115 V UV Lamp and Base Stand 21 in. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
17195080
View Documents Promotion Details
Labconco
5340810
Stainless Steel 230 V UV Lamp and Base Stand 21 in. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
17195029
View Documents Promotion Details
Labconco
5341000
Epoxy-coated Steel 115 V Clean Bench Only 26 in. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
17194957
View Documents Promotion Details
Labconco
5341010
Epoxy-coated Steel 230 V Clean Bench Only 26 in. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
17194963
View Documents Promotion Details
Labconco
5341100
Stainless Steel 115 V Clean Bench Only 26 in. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
17195026
View Documents Promotion Details
Labconco
5341110
Stainless Steel 230 V Clean Bench Only 26 in. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
17194958
View Documents Promotion Details
Labconco
5341200
Epoxy-coated Steel 115 V UV Lamp 26 in. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
17195097
View Documents Promotion Details
Labconco
5341210
Epoxy-coated Steel 230 V UV Lamp 26 in. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
17195061
View Documents Promotion Details
Labconco
5341300
Stainless Steel 115 V UV Lamp 26 in. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
17195038
View Documents Promotion Details
Labconco
5341310
Stainless Steel 230 V UV Lamp 26 in. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
17195067
View Documents Promotion Details
Labconco
5341500
Epoxy-coated Steel 115 V Base Stand 26 in. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
17195062
View Documents Promotion Details
Labconco
5341510
Epoxy-coated Steel 230 V Base Stand 26 in. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
17195035
View Documents Promotion Details
Labconco
5341600
Stainless Steel 115 V Base Stand 26 in. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
17194945
View Documents Promotion Details
Labconco
5341610
Stainless Steel 230 V Base Stand 26 in. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
17195045
View Documents Promotion Details
Labconco
5341700
Epoxy-coated Steel 115 V UV Lamp and Base Stand 26 in. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
17195047
View Documents Promotion Details
Labconco
5341710
Epoxy-coated Steel 230 V UV Lamp and Base Stand 26 in. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
17195063
View Documents Promotion Details
Labconco
LCN5341800
Stainless Steel 115 V UV Lamp and Base Stand 26 in. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
17195033
View Documents Promotion Details
Labconco
5341810
Stainless Steel 230 V Base Stand, UV Lamp 26 in. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

  • 4 ft. Nexus Horizontal Clean Bench
  • Internal work area
  • ISO Class 4 laminar airflow is 10 times cleaner than the industry standard ISO Class 5
  • Energy efficient ECM motor with CAP™ Technology ensures accurate airflow between certifications
Specifications

Specifications

Horizontal Clean Bench
HEPA
50.1 in.
127.5 cm
None
51.4 in.
130.6 cm
Epoxy
Optic White
30.2 in.
76.7 cm
NEMA 5-15P
49.0 in.
124.5 cm
Videos
SDS
Documents

Documents