missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco 4 ft. Nexus Horizontal Clean Bench
Nexus Horizontal Clean Benches are designed to protect non-hazardous applications by providing an ISO Class 4 clean airflow environment. The large, 5 in. touchscreen interface features Nexus OS™ software that simplifies operation, maintains records and provides user customizaton of the system.
Supplier: Labconco 5341710
Description
- 4 ft. Nexus Horizontal Clean Bench
- Internal work area
- ISO Class 4 laminar airflow is 10 times cleaner than the industry standard ISO Class 5
- Energy efficient ECM motor with CAP™ Technology ensures accurate airflow between certifications
Chemical Identifiers
| Horizontal Clean Bench | |
| HEPA | |
| 50.1 in. | |
| 127.5 cm | |
| None | |
| 51.4 in. | |
| 130.6 cm |
| Epoxy | |
| Optic White | |
| 30.2 in. | |
| 76.7 cm | |
| NEMA 5-15P | |
| 49.0 in. | |
| 124.5 cm |
Specifications
| Horizontal Clean Bench | |
| HEPA | |
| Epoxy-coated Steel | |
| 26 in. | |
| 50.1 in. | |
| 127.5 cm | |
| UV Lamp and Base Stand | |
| NEMA 5-15P | |
| 51.4 in. | |
| 130.6 cm |
| Epoxy | |
| UV | |
| Optic White | |
| 66.6 cm | |
| 30.2 in. | |
| 76.7 cm | |
| None | |
| 230 V | |
| 49.0 in. | |
| 124.5 cm |