missing translation for 'onlineSavingsMsg'
Learn More

4-Formylphenoxyacetic acid, 98%

Catalog No. AAA1396030
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1396030 250 g
AAA1396009 10 g
AAA1396018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1396030 Supplier Thermo Scientific Chemicals Supplier No. A1396030

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 22042-71-3 | C9H7O4 | 179.15 g/mol

4-Formylphenoxyacetic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Formylphenoxyacetic acid is used as an organic chemical synthesis intermediate.

Solubility
Soluble in hot methanol within almost transparency.

Notes
Air sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 22042-71-3
Molecular Formula C9H7O4
Molecular Weight (g/mol) 179.15
MDL Number MFCD00016613
InChI Key OYNIIKHNXNPSAG-UHFFFAOYSA-M
Synonym 4-formylphenoxyacetic acid, 4-formylphenoxy acetic acid, 2-4-formylphenoxy acetic acid, p-formylphenoxyacetic acid, acetic acid, 4-formylphenoxy, acetic acid, 2-4-formylphenoxy, p-formylphenoxy acetic acid, 4-carboxymethyloxy benzaldehyde, 4-formyl phenoxy acetic acid, 4-formylphenoxyactic acid
PubChem CID 89177
IUPAC Name 2-(4-formylphenoxy)acetic acid
SMILES [O-]C(=O)COC1=CC=C(C=O)C=C1

Specifications

Melting Point 193°C to 198°C
Quantity 250 g
Beilstein 779885
Sensitivity Air Sensitive
Solubility Information Soluble in hot methanol within almost transparency.
Formula Weight 180.16
Percent Purity 98%
Chemical Name or Material 4-Formylphenoxyacetic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 244-749-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only