CAS RN 22042-71-3
IUPAC Name:
2-(4-formylphenoxy)acetate
Synonyms:
2-(4-formylphenoxy)acetate
Molecular Weight (g/mol):
179.15
Molecular Formula:
C9H7O4
InChi Key:
OYNIIKHNXNPSAG-UHFFFAOYSA-M
SMILES:
[O-]C(=O)COC1=CC=C(C=O)C=C1
1
–
1
of
1
results
1
Promotions Available
4-Formylphenoxyacetic acid, 98%
4-Formylphenoxyacetic acid, 98%
CAS: 22042-71-3 Molecular Formula: C9H7O4 Molecular Weight (g/mol): 179.15 MDL Number: MFCD00016613 InChI Key: OYNIIKHNXNPSAG-UHFFFAOYSA-M Synonym: 4-formylphenoxyacetic acid,4-formylphenoxy acetic acid,2-4-formylphenoxy acetic acid,p-formylphenoxyacetic acid,acetic acid, 4-formylphenoxy,acetic acid, 2-4-formylphenoxy,p-formylphenoxy acetic acid,4-carboxymethyloxy benzaldehyde,4-formyl phenoxy acetic acid,4-formylphenoxyactic acid PubChem CID: 89177 IUPAC Name: 2-(4-formylphenoxy)acetic acid SMILES: [O-]C(=O)COC1=CC=C(C=O)C=C1