missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Formyl-trans-stilbene 98.0+%, TCI America™
$227.41 - $721.40
Chemical Identifiers
| CAS | 40200-69-9 |
|---|---|
| Molecular Formula | C15H12O |
| Molecular Weight (g/mol) | 208.26 |
| MDL Number | MFCD00011580 |
| InChI Key | CLXSBHRRZNBTRT-VOTSOKGWSA-N |
| Synonym | trans-4-Stilbenecarboxaldehyde |
| PubChem CID | 5375876 |
| IUPAC Name | 4-[(E)-2-phenylethenyl]benzaldehyde |
| SMILES | C1=CC=C(C=C1)C=CC2=CC=C(C=C2)C=O |
Chemical Identifiers
| 40200-69-9 | |
| 208.26 | |
| CLXSBHRRZNBTRT-VOTSOKGWSA-N | |
| 5375876 | |
| C1=CC=C(C=C1)C=CC2=CC=C(C=C2)C=O |
| C15H12O | |
| MFCD00011580 | |
| trans-4-Stilbenecarboxaldehyde | |
| 4-[(E)-2-phenylethenyl]benzaldehyde |
Specifications
| 40200-69-9 | |
| Yellow | |
| MFCD00011580 | |
| trans-4-Stilbenecarboxaldehyde | |
| C1=CC=C(C=C1)C=CC2=CC=C(C=C2)C=O | |
| 208.26 | |
| 208.26 | |
| Crystalline Powder |
| 114°C | |
| C15H12O | |
| 5 g | |
| CLXSBHRRZNBTRT-VOTSOKGWSA-N | |
| 4-[(E)-2-phenylethenyl]benzaldehyde | |
| 5375876 | |
| ≥98.0% (GC) | |
| 4-Formyl-trans-stilbene |
Safety and Handling
TSCA : No