accessibility menu, dialog, popup

UserName

CAS RN 40200-69-9

O(E/Z)

CAS RN 40200-69-9

IUPAC Name: 4-(2-phenylethenyl)benzaldehyde
Synonyms: 4-(2-phenylethenyl)benzaldehyde
Molecular Weight (g/mol): 208.26
Molecular Formula: C15H12O
InChi Key: CLXSBHRRZNBTRT-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(C=CC2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results